missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PAIP1 Full-length ORF (NP_877590.1, 1 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_877590.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10605 |
Molecular Weight (g/mol) | 72kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16145402
|
Abnova™
H00010605-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16135402
|
Abnova™
H00010605-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified. [provided by RefSeq]
Sequence: MSDGFDRAPEQTRPLRAPPSSQDKIPQQNSESAMAKPQVVVAPVLMSKLSVNAPEFYPSGYSSSYTESYEDGCEDYPTLSEYVQDFLNHLTEQPGSFETEIEQFAETLNGCVTTDDALQELVELIYQQATSIPNFSYMGARLCNYLSHHLTISPQSGNFRQLLLQRCRTEYEVKDQAAKGDEVTRKRFHAFVLFLGELYLNLEIKGTNGQVTRADILQVGLRELLNALFSNPMDDNLICAVKLLKLTGSVLEDAWKEKGKMDMEEIIQRIENVVLDANCSRDVKQMLLKLVELRSSNWGRVHATSTYREATPENDPNYFMNEPTFYTSDGVPFTAADPDYQEKYQELLEREDFFPDYEENGTDLSGAGDPYLDDIDDEMDPEIEEAYEKFCLESERKRKQSpecifications
NP_877590.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSDGFDRAPEQTRPLRAPPSSQDKIPQQNSESAMAKPQVVVAPVLMSKLSVNAPEFYPSGYSSSYTESYEDGCEDYPTLSEYVQDFLNHLTEQPGSFETEIEQFAETLNGCVTTDDALQELVELIYQQATSIPNFSYMGARLCNYLSHHLTISPQSGNFRQLLLQRCRTEYEVKDQAAKGDEVTRKRFHAFVLFLGELYLNLEIKGTNGQVTRADILQVGLRELLNALFSNPMDDNLICAVKLLKLTGSVLEDAWKEKGKMDMEEIIQRIENVVLDANCSRDVKQMLLKLVELRSSNWGRVHATSTYREATPENDPNYFMNEPTFYTSDGVPFTAADPDYQEKYQELLEREDFFPDYEENGTDLSGAGDPYLDDIDDEMDPEIEEAYEKFCLESERKRKQ | |
RUO | |
PAIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10605 | |
PAIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12360 | |
PAIP1 | |
Recombinant | |
wheat germ expression system |