Learn More
Abnova™ Human PANK2 Partial ORF (NP_705902.2, 437 a.a. - 570 a.a.) Recombinant Protein with GST-tag at N-terminal
Liquid
Brand: Abnova™ H00080025-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein belonging to the pantothenate kinase family and is the only member of that family to be expressed in mitochondria. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by acyl CoA species. Mutations in this gene are associated with HARP syndrome and pantothenate kinase-associated neurodegeneration (PKAN), formerly Hallervorden-Spatz syndrome. Alternative splicing, involving the use of alternate first exons, results in multiple transcripts encoding different isoforms. [provided by RefSeq]
Sequence: MASRGDSTKVDKLVRDIYGGDYERFGLPGWAVASSFGNMMSKEKREAVSKEDLARATLITITNNIGSIARMCALNENINQVVFVGNFLRINTIAMRLLAYALDYWSKGQLKALFSEHEGYFGAVGALLELLKIPSpecifications
NP_705902.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
41.2kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MASRGDSTKVDKLVRDIYGGDYERFGLPGWAVASSFGNMMSKEKREAVSKEDLARATLITITNNIGSIARMCALNENINQVVFVGNFLRINTIAMRLLAYALDYWSKGQLKALFSEHEGYFGAVGALLELLKIP | |
RUO | |
PANK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80025 | |
PANK2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf48/FLJ17232/HARP/HSS/MGC15053/NBIA1/PKAN | |
PANK2 | |
Recombinant | |
wheat germ expression system |