Learn More
Abnova™ Human PANX2 Partial ORF (NP_443071.1, 133 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_443071.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56666 |
Molecular Weight (g/mol) | 35.31kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188786
|
Abnova™
H00056666-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178786
|
Abnova™
H00056666-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the innexin family. Innexin family members are the structural components of gap junctions. This protein and pannexin 1 are abundantly expressed in central nervous system (CNS) and are coexpressed in various neuronal populations. Studies in Xenopus oocytes suggest that this protein alone and in combination with pannexin 1 may form cell type-specific gap junctions with distinct properties. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: EFLASTRLTSELNFLLQEIDNCYHRAAEGRAPKIEKQIQSKGPGITEREKREIIENAEKEKSPEQNLFEKYLERRGRSNFLAKLYLASpecifications
NP_443071.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119432/PX2/hPANX2 | |
PANX2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
56666 | |
PANX2 (Human) Recombinant Protein (Q01) | |
EFLASTRLTSELNFLLQEIDNCYHRAAEGRAPKIEKQIQSKGPGITEREKREIIENAEKEKSPEQNLFEKYLERRGRSNFLAKLYLA | |
RUO | |
PANX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |