Learn More
Abnova™ Human PANX2 Partial ORF (NP_443071.1, 133 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056666-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the innexin family. Innexin family members are the structural components of gap junctions. This protein and pannexin 1 are abundantly expressed in central nervous system (CNS) and are coexpressed in various neuronal populations. Studies in Xenopus oocytes suggest that this protein alone and in combination with pannexin 1 may form cell type-specific gap junctions with distinct properties. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: EFLASTRLTSELNFLLQEIDNCYHRAAEGRAPKIEKQIQSKGPGITEREKREIIENAEKEKSPEQNLFEKYLERRGRSNFLAKLYLASpecifications
NP_443071.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.31kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EFLASTRLTSELNFLLQEIDNCYHRAAEGRAPKIEKQIQSKGPGITEREKREIIENAEKEKSPEQNLFEKYLERRGRSNFLAKLYLA | |
RUO | |
PANX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56666 | |
PANX2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119432/PX2/hPANX2 | |
PANX2 | |
Recombinant | |
wheat germ expression system |