missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PAPSS1 Full-length ORF (AAH11392.1, 1 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH11392.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9061 |
Molecular Weight (g/mol) | 94.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16183232
|
Abnova™
H00009061-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16193232
|
Abnova™
H00009061-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 [PubMed 10679223]). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2 (MIM 603005).[supplied by OMIM]
Sequence: MQRATNVTYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTVSMALEEYLVCHGIPCYTLDGDNIRQGLNKNLGFSPEDREENVRRIAEVAKLFADAGLVCITSFISPYTQDRNNARQIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAETLPALKINKVDMQWVQVLAEGWATPLNGFMREREYLQCLHFDCLLDGGVINLSVPIVLTATHEDKERLDGCTAFALMYEGRRVAILRNPEFFEHRKEERCARQWGTTCKNHPYIKMVMEQGDWLIGGDLQVLDRVYWNDGLDQYRLTPTELKQKFKDMNADAVFAFQLRNPVHNGHALLMQDTHKQLLERGYRRPVLLLHPLGGWTKDDDVPLMWRMKQHAAVLEEGVLNPETTVVAIFPSPMMYAGPTEVQWHCRARMVAGANFYIVGRDPAGMPHPETGKDLYEPSHGAKVLTMAPGLITLEIVPFRVAAYNKKKKRMDYYDSEHHEDFEFISGTRMRKLAREGQKPPEGFMAPKAWTVLTEYYKSLEKASpecifications
AAH11392.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
94.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQRATNVTYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTVSMALEEYLVCHGIPCYTLDGDNIRQGLNKNLGFSPEDREENVRRIAEVAKLFADAGLVCITSFISPYTQDRNNARQIHEGASLPFFEVFVDAPLHVCEQRDVKGLYKKARAGEIKGFTGIDSEYEKPEAPELVLKTDSCDVNDCVQQVVELLQERDIVPVDASYEVKELYVPENKLHLAKTDAETLPALKINKVDMQWVQVLAEGWATPLNGFMREREYLQCLHFDCLLDGGVINLSVPIVLTATHEDKERLDGCTAFALMYEGRRVAILRNPEFFEHRKEERCARQWGTTCKNHPYIKMVMEQGDWLIGGDLQVLDRVYWNDGLDQYRLTPTELKQKFKDMNADAVFAFQLRNPVHNGHALLMQDTHKQLLERGYRRPVLLLHPLGGWTKDDDVPLMWRMKQHAAVLEEGVLNPETTVVAIFPSPMMYAGPTEVQWHCRARMVAGANFYIVGRDPAGMPHPETGKDLYEPSHGAKVLTMAPGLITLEIVPFRVAAYNKKKKRMDYYDSEHHEDFEFISGTRMRKLAREGQKPPEGFMAPKAWTVLTEYYKSLEKA | |
RUO | |
PAPSS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9061 | |
PAPSS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATPSK1/PAPSS/SK1 | |
PAPSS1 | |
Recombinant | |
wheat germ expression system |