Learn More
Abnova™ Human PAPSS2 Partial ORF (NP_004661, 513 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004661 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9060 |
Molecular Weight (g/mol) | 36.74kDa |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
16169835
|
Abnova™
H00009060-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16179835
|
Abnova™
H00009060-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq]
Sequence: DPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLESpezifikation
NP_004661 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATPSK2/SK2 | |
PAPSS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9060 | |
PAPSS2 (Human) Recombinant Protein (Q01) | |
DPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLE | |
RUO | |
PAPSS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |