Learn More
Abnova™ Human PAX9 Full-length ORF (NP_006185.1, 1 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005083-P02.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements. [provided by RefSeq]
Sequence: MEPAFGEVNQLGGVFVNGRPLPNAIRLRIVELAQLGIRPCDISRQLRVSHGCVSKILARYNETGSILPGAIGGSKPRVTTPTVVKHIRTYKQRDPGIFAWEIRDRLLADGVCDKYNVPSVSSISRILRNKIGNLAQQGHYDSYKQHQPTPQPALPYNHIYSYPSPITAAAAKVPTPPGVPAIPGSVAMPRTWPSSHSVTDILGIRSITDQVSDSSPYHSPKVEEWSSLGRNNFPAAAPHAVNGLEKGALEQEAKYGQAPNGLPAVGSFVSASSMAPYPTPAQVSPYMTYSAAPSGYVAGHGWQHAGGTSLSPHNCDIPASLAFKGMQAAREGSHSVTASALSpecifications
NP_006185.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAX9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5083 | |
PAX9 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPAFGEVNQLGGVFVNGRPLPNAIRLRIVELAQLGIRPCDISRQLRVSHGCVSKILARYNETGSILPGAIGGSKPRVTTPTVVKHIRTYKQRDPGIFAWEIRDRLLADGVCDKYNVPSVSSISRILRNKIGNLAQQGHYDSYKQHQPTPQPALPYNHIYSYPSPITAAAAKVPTPPGVPAIPGSVAMPRTWPSSHSVTDILGIRSITDQVSDSSPYHSPKVEEWSSLGRNNFPAAAPHAVNGLEKGALEQEAKYGQAPNGLPAVGSFVSASSMAPYPTPAQVSPYMTYSAAPSGYVAGHGWQHAGGTSLSPHNCDIPASLAFKGMQAAREGSHSVTASAL | |
RUO | |
PAX9 | |
Recombinant | |
wheat germ expression system |