Learn More
Abnova™ Human PCDHA6 Partial ORF (NP_061732, 295 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056142-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. [provided by RefSeq]
Sequence: IDRNTGEIVIRGNLDFEQENLYKILIDATDKGHPPMAGHCTVLVRILDKNDNVPEIALTSLSLPVREDAQFGTVIASpecifications
NP_061732 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IDRNTGEIVIRGNLDFEQENLYKILIDATDKGHPPMAGHCTVLVRILDKNDNVPEIALTSLSLPVREDAQFGTVIA | |
RUO | |
PCDHA6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56142 | |
PCDHA6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNR2/CNRN2/CNRS2/CRNR2/PCDH-ALPHA6 | |
PCDHA6 | |
Recombinant | |
wheat germ expression system |