Learn More
Abnova™ Human PCDHB6 Full-length ORF (AAI52976.1, 1 a.a. - 794 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAI52976.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56130 |
Molecular Weight (g/mol) | 114.29kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172093
|
Abnova™
H00056130-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182093
|
Abnova™
H00056130-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]
Sequence: MMQTKVQNKKRQVAFFILLMLWGEVGSESIQYSVLEETESGTFVANLTKDLGLRVGELASRGARVVFKGNRQHLQFDPQTHDLLLNEKLDREELCGSTEPCVLPFQVLLENPLQFFQASLRVRDINDHAPEFPAREMLLKISEITMPGKIFPLKMAHDLDTGSNGLQRYTISSNPHFHVLTRNRSEGRKFPELVLDKPLDREEQPQLRLTLIALDGGSPPRSGTSEIQIQVLDINDNVPEFAQELYEAQVPENNPLGSLVITVSARDLDAGSFGKVSYALFQVDDVNQPFEINAITGEIRLRKALDFEEIQSYDVDVEATDGGGLSGKCSLVVRVLDVNDNAPELTMSFFISLIPENLPEITVAVFSVSDADSGHNQQVICSIENNLPFLLRPSVENFYTLVTEGALDRESRAEYNITITVTDLGTPRLKTQQSITVQVSDVNDNVPAFTQTSYTLFVRENNSPALHIGSVSATDRDSGINAQVTYSLLPPQDPHLPLSSLVSINADNGHLFALRSLDYEALQSFEFRVGATDRGSPALSSEALVRLLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATELGLFGVWAHNGEVRTARLLSERDAAKHRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRYSVPEGPFPGHLVDVSGTGTLSQSYQYKVCLTGGSETNEFKFLKPIMPNFPPQGTEREMEETPTSRNSFPFSSpecifications
AAI52976.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114.29kDa | |
Glutathione Sepharose 4 Fast Flow | |
MMQTKVQNKKRQVAFFILLMLWGEVGSESIQYSVLEETESGTFVANLTKDLGLRVGELASRGARVVFKGNRQHLQFDPQTHDLLLNEKLDREELCGSTEPCVLPFQVLLENPLQFFQASLRVRDINDHAPEFPAREMLLKISEITMPGKIFPLKMAHDLDTGSNGLQRYTISSNPHFHVLTRNRSEGRKFPELVLDKPLDREEQPQLRLTLIALDGGSPPRSGTSEIQIQVLDINDNVPEFAQELYEAQVPENNPLGSLVITVSARDLDAGSFGKVSYALFQVDDVNQPFEINAITGEIRLRKALDFEEIQSYDVDVEATDGGGLSGKCSLVVRVLDVNDNAPELTMSFFISLIPENLPEITVAVFSVSDADSGHNQQVICSIENNLPFLLRPSVENFYTLVTEGALDRESRAEYNITITVTDLGTPRLKTQQSITVQVSDVNDNVPAFTQTSYTLFVRENNSPALHIGSVSATDRDSGINAQVTYSLLPPQDPHLPLSSLVSINADNGHLFALRSLDYEALQSFEFRVGATDRGSPALSSEALVRLLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATELGLFGVWAHNGEVRTARLLSERDAAKHRLVVLVKDNGEPPRSATATLHVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRYSVPEGPFPGHLVDVSGTGTLSQSYQYKVCLTGGSETNEFKFLKPIMPNFPPQGTEREMEETPTSRNSFPFS | |
RUO | |
PCDHB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56130 | |
PCDHB6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCDH-BETA6 | |
PCDHB6 | |
Recombinant | |
wheat germ expression system |