missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDHB7 Partial ORF (NP_061763, 90 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056129-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132093

  • 4515.00 SEK / 10µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. The transcript for this particular family member uses more than one polyadenylation site. [provided by RefSeq]

Sequence: LDREELCGPREPCVLPFQLLLEKPFQIFRAELWVRDINDHAPVFLDREISLKILESTTPGAAFLLESAQDSDVGTNSLSNYTISPNAYFHINVHDSGEGN
Specifications

Specifications

NP_061763
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LDREELCGPREPCVLPFQLLLEKPFQIFRAELWVRDINDHAPVFLDREISLKILESTTPGAAFLLESAQDSDVGTNSLSNYTISPNAYFHINVHDSGEGN
RUO
PCDHB7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56129
PCDHB7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111391/MGC163205/PCDH-BETA7
PCDHB7
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers