Learn More
Abnova™ Human PCSK7 Partial ORF (NP_004707, 434 a.a. - 543 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004707 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9159 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130086
|
Abnova™
H00009159-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16120086
|
Abnova™
H00009159-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a calcium-dependent serine endoprotease. It is structurally related to its family members, PACE and PACE4. This protein is concentrated in the trans-Golgi network, associated with the membranes, and is not secreted. It can process proalbumin and is thought to be responsible for the activation of HIV envelope glycoproteins gp160 and gp140. This gene has been implicated in the transcriptional regulation of housekeeping genes. Multiple alternatively spliced transcripts are described for this gene but their full length nature is not yet known. Downstream of this gene's map location at 11q23-q24, nucleotides that match part of this gene's 3' end are duplicated and inverted. A translocation breakpoint associated with lymphoma occurs between this gene and its inverted counterpart. [provided by RefSeq]
Sequence: IIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLESpecifications
NP_004707 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LPC/PC7/PC8/SPC7 | |
PCSK7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9159 | |
PCSK7 (Human) Recombinant Protein (Q01) | |
IIVFTATRYEDRRAEWVTNEAGFSHSHQHGFGLLNAWRLVNAAKIWTSVPYLASYVSPVLKENKAIPQSPRSLEVLWNVSRMDLEMSGLKTLEHVAVTVSITHPRRGSLE | |
RUO | |
PCSK7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |