missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PDHA2 Partial ORF (NP_005381.1, 7 a.a. - 115 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005161-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16182555

  • 4515.00 SEK / 10µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: SRVLRRVAQKSARRVLVASRNSSNDATFEIKKCDLYLLEEGPPVTTVLTRAEGLKYYRMMLTVRRMELKADQLYKQKFIRGFCHLCDGQEACCVGLEAGINPSDHVITS
Specifications

Specifications

NP_005381.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SRVLRRVAQKSARRVLVASRNSSNDATFEIKKCDLYLLEEGPPVTTVLTRAEGLKYYRMMLTVRRMELKADQLYKQKFIRGFCHLCDGQEACCVGLEAGINPSDHVITS
RUO
PDHA2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5161
PDHA2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC149517/MGC149518/PDHAL
PDHA2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers