Learn More
Abnova™ Human PDHX Partial ORF (NP_003468, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008050-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The PDHX gene encodes component X of the pyruvate dehydrogenase (PDH) complex. For a detailed description of the pyruvate dehydrogenase complex, see MIM 300502. The mammalian PDH complex differs from that in E. coli and from the other mammalian alpha-keto acid dehydrogenases by the presence of a 53-kD protein called protein X. Component X binds to the E3 (MIM 238331) component of the PDH complex (Robinson et al., 1990 [PubMed 2112155]; Aral et al., 1997 [PubMed 9399911]).[supplied by OMIM]
Sequence: MAASWRLGCDPRLLRYLVGFPGRRSVGLVKGALGWSVSRGRNWRWFHSTQWLRGDPIKILMPSLSPTMEEGNIVKWLKKEGEAVSAGDALCEIETDKAVVTLDASDDGISpecifications
NP_003468 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAASWRLGCDPRLLRYLVGFPGRRSVGLVKGALGWSVSRGRNWRWFHSTQWLRGDPIKILMPSLSPTMEEGNIVKWLKKEGEAVSAGDALCEIETDKAVVTLDASDDGI | |
RUO | |
PDHX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8050 | |
PDHX (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DLDBP/E3BP/OPDX/PDX1/proX | |
PDHX | |
Recombinant | |
wheat germ expression system |