missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PELP1 Partial ORF (AAW80659.1, 337 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAW80659.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27043 |
Molecular Weight (g/mol) | 33.77kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16145656
|
Abnova™
H00027043-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16135656
|
Abnova™
H00027043-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PELP1 is a coactivator of estrogen receptor (see ESR1; MIM 133430)-mediated transcription and a corepressor of other nuclear hormone receptors and sequence-specific transcription factors (Choi et al., 2004 [PubMed 15456770]).[supplied by OMIM]
Sequence: LNSWSIGRDSLSPGQERPYSTVRTKVYAILELWVQVCGASAGMLQGGASGEALLTHLLSDISPPADALKLRSPRSpecifications
AAW80659.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HMX3/MNAR/P160 | |
PELP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
27043 | |
PELP1 (Human) Recombinant Protein (Q01) | |
LNSWSIGRDSLSPGQERPYSTVRTKVYAILELWVQVCGASAGMLQGGASGEALLTHLLSDISPPADALKLRSPR | |
RUO | |
PELP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |