missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Peroxiredoxin 3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7504
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peroxiredoxin 3 Monoclonal specifically detects Peroxiredoxin 3 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifikationer
| Peroxiredoxin 3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P30048 | |
| PRDX3 | |
| E. coli-derived recombinant human Peroxiredoxin 3 Met1-Gln256 Accession # P30048 | |
| 100 μg | |
| Primary | |
| Detects human Peroxiredoxin 3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Peroxiredoxin 1, 2, 4, or recombinant mouse Peroxiredoxin 3 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 774406 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antioxidant protein 1MGC104387, AOP-1, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, PRDX3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10935 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering