Learn More
Abnova™ Human PEX6 Full-length ORF (AAH48331.1, 1 a.a. - 980 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005190-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6. [provided by RefSeq]
Sequence: MALAVLRVLEPFPTETPPLAVLLPPGGPWPAAELGLVLALRPAGESPAGPALLVAALEGPDAGTEEQGPGPPQLLVSRALLRLLALGSGAWVRARAVRRPPALGWALLGTSLGPGLGPRVGPLLVRRGETLPVPGPRVLETRPALQGLLGPGTRLAVTELRGRARLCPESGDSSRPPPPPVVSSFAVSGTVRRLQGVLGGTGDSLGVSRSCLRGLGLFQGEWVWVAQARESSNTSQPHLARVQVLEPRWDLSDRLGPGSGPLGEPLADGLALVPATLAFNLGCDPLEMGELRIQRYLEGSIAPEDKGSCSLLPGPPFARELHIEIVSSPHYSTNGNYDGVLYRHFQIPRVVQEGDVLCVPTIGQVEILEGSPEKLPRWREMFFKVKKTVGEAPDGPASAYLADTTHTSLYMVGSTLSPVPWLPSEESTLWSSLSPPGLEALVSELCAVLKPRLQPGGALLTGTSSVLLRGPPGCGKTTVVAAACSHLGLHLLKVPCSSLCAESSGAVETKLQAIFSRARRCRPAVLLLTAVDLLGRDRDGLGEDARVMAVLRHLLLNEDPLNSCPPLMVVATTSRAQDLPADVQTAFPHELEVPALSEGQRLSILRALTAHLPLGQEVNLAQLARRCAGFVVGDLYALLTHSSRAACTRIKNSGLAGGLTEEDEGELCAAGFPLLAEDFGQALEQLQTAHSQAVGAPKIPSVSWHDVGGLQEVKKEILETIQLPLEHPELLSLGLRRSGLLLHGPPGTGKTLLAKAVATECSLTFLSVKGPELINMYVGQSEENVREVFARARAAAPCIIFFDELDSLAPSRGRSGDSGGVMDRVVSQLLAELDGLHSTQDVFVIGATNRPDLLDPALLRPGRFDKLVFVGANEDRASQLRVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAACSpecifications
AAH48331.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
130.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAF-2/PAF2/PXAAA1 | |
PEX6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5190 | |
PEX6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALAVLRVLEPFPTETPPLAVLLPPGGPWPAAELGLVLALRPAGESPAGPALLVAALEGPDAGTEEQGPGPPQLLVSRALLRLLALGSGAWVRARAVRRPPALGWALLGTSLGPGLGPRVGPLLVRRGETLPVPGPRVLETRPALQGLLGPGTRLAVTELRGRARLCPESGDSSRPPPPPVVSSFAVSGTVRRLQGVLGGTGDSLGVSRSCLRGLGLFQGEWVWVAQARESSNTSQPHLARVQVLEPRWDLSDRLGPGSGPLGEPLADGLALVPATLAFNLGCDPLEMGELRIQRYLEGSIAPEDKGSCSLLPGPPFARELHIEIVSSPHYSTNGNYDGVLYRHFQIPRVVQEGDVLCVPTIGQVEILEGSPEKLPRWREMFFKVKKTVGEAPDGPASAYLADTTHTSLYMVGSTLSPVPWLPSEESTLWSSLSPPGLEALVSELCAVLKPRLQPGGALLTGTSSVLLRGPPGCGKTTVVAAACSHLGLHLLKVPCSSLCAESSGAVETKLQAIFSRARRCRPAVLLLTAVDLLGRDRDGLGEDARVMAVLRHLLLNEDPLNSCPPLMVVATTSRAQDLPADVQTAFPHELEVPALSEGQRLSILRALTAHLPLGQEVNLAQLARRCAGFVVGDLYALLTHSSRAACTRIKNSGLAGGLTEEDEGELCAAGFPLLAEDFGQALEQLQTAHSQAVGAPKIPSVSWHDVGGLQEVKKEILETIQLPLEHPELLSLGLRRSGLLLHGPPGTGKTLLAKAVATECSLTFLSVKGPELINMYVGQSEENVREVFARARAAAPCIIFFDELDSLAPSRGRSGDSGGVMDRVVSQLLAELDGLHSTQDVFVIGATNRPDLLDPALLRPGRFDKLVFVGANEDRASQLRVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAAC | |
RUO | |
PEX6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |