Learn More
Novus Biologicals™ Human PGAM2 ELISA Kit (Colorimetric)
This kit recognizes Human PGAM2 in samples.
Brand:  Novus Biologicals™ NBP2-81248
Product Code. 18323800
  • 7482.53 SEK / 1 set
Explore more special offers
Description
Description
No significant cross-reactivity or interference between Human PGAM2 and analogues was observed.
Specifications
Specifications
0.16 to 10 ng/mL
ELISA Kit (Colorimetric)
ELISA
PGAM2
1 Kit
Storage of components varies. See protocol for specific instructions.
0.09 ng/mL
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
5224
Human
Cancer, Proteases & Other Enzymes, Signal Transduction
PGAM2