Learn More
Novus Biologicals™ Human PGAM2 ELISA Kit (Colorimetric)
This kit recognizes Human PGAM2 in samples.
Brand: Novus Biologicals™ NBP2-81248
This item is not returnable.
View return policy
Description
No significant cross-reactivity or interference between Human PGAM2 and analogues was observed.Specifications
0.16 to 10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
PGAM2 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |
0.09 ng/mL | |
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum | |
5224 | |
Human | |
Cancer, Proteases & Other Enzymes, Signal Transduction | |
PGAM2 |