missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PGF Partial ORF (AAH01422, 27 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005228-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: WALSAGNGSSEVEVVPFQEVWGRSYCRALERLVDVVSEYPSEVEHMFSPSCVSLLRCTGCCGDENLHCVPVETANVTMQLLKIRSGDRPSYVELTFSQHVSpecifications
AAH01422 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WALSAGNGSSEVEVVPFQEVWGRSYCRALERLVDVVSEYPSEVEHMFSPSCVSLLRCTGCCGDENLHCVPVETANVTMQLLKIRSGDRPSYVELTFSQHV | |
RUO | |
PGF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5228 | |
PGF (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D12S1900/PGFL/PLGF/PlGF-2/SHGC-10760 | |
PGF | |
Recombinant | |
wheat germ expression system |