missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PGRMC1 Full-length ORF (NP_006658.1, 1 a.a. - 195 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010857-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135762

  • 5030.41 SEK / 25µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Progesterone binding protein is a putative steroid membrane receptor. The protein is expressed predominantly in the liver and kidney. [provided by RefSeq]

Sequence: MAAEDVVATGADPSDLESGGLLHEIFTSPLNLLLLGLCIFLLYKIVRGDQPAASGDSDDDEPPPLPRLKRRDFTPAELRRFDGVQDPRILMAINGKVFDVTKGRKFYGPEGPYGVFAGRDASRGLATFCLDKEALKDEYDDLSDLTAAQQETLSDWESQFTFKYHHVGKLLKEGEEPTVYSDEEEPKDESARKND
Specifications

Specifications

NP_006658.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPR6.6/MPR
PGRMC1
Yes
wheat germ expression system
Liquid
10857
PGRMC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAEDVVATGADPSDLESGGLLHEIFTSPLNLLLLGLCIFLLYKIVRGDQPAASGDSDDDEPPPLPRLKRRDFTPAELRRFDGVQDPRILMAINGKVFDVTKGRKFYGPEGPYGVFAGRDASRGLATFCLDKEALKDEYDDLSDLTAAQQETLSDWESQFTFKYHHVGKLLKEGEEPTVYSDEEEPKDESARKND
RUO
PGRMC1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates