missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PGS1 Full-length ORF (ENSP00000262764, 1 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009489-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MKGQIRVAKRRVVMASLYLGTGPLEQELVDCLESTLEKSLQAKFPSNLKVSILLDFTRGSRGRKNSRTMLLPLLRRFPEQVRVSLFHTPHLRGLLRLLIPERFNETIGLQHIKVYLFDNSVILSGANLSDSYFTNRQDRYVFLQDCAEIADFFTELVDAVGDVSLQLQGDDTVQVVDGMVHPYKGDRAEYCKAANKRVMDVINSARTRQQMLHAQTFHSNSLLTQEDAAAAGDRRPAPDTWIYPLIQMKPFEIQIDEIVTETLLTEAERGAKVYLTTGYFNLTQAYMDLVLGTRAEYQILLASPEVNGFFGAKGVAGAIPAAYVHIERQFFSEVCSLGQQERVQLQEYWRRGWTFHAKGLWLYLAGSSLPCLTLIGSPNFGYRSVHRDLEAQIAIVTENQALQQQLHQEQEQLYLRSGVVSSATFEQPSRQVKLWVKMVTPLIKNFFSpecifications
ENSP00000262764 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp762M186/MGC131960 | |
PGS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9489 | |
PGS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKGQIRVAKRRVVMASLYLGTGPLEQELVDCLESTLEKSLQAKFPSNLKVSILLDFTRGSRGRKNSRTMLLPLLRRFPEQVRVSLFHTPHLRGLLRLLIPERFNETIGLQHIKVYLFDNSVILSGANLSDSYFTNRQDRYVFLQDCAEIADFFTELVDAVGDVSLQLQGDDTVQVVDGMVHPYKGDRAEYCKAANKRVMDVINSARTRQQMLHAQTFHSNSLLTQEDAAAAGDRRPAPDTWIYPLIQMKPFEIQIDEIVTETLLTEAERGAKVYLTTGYFNLTQAYMDLVLGTRAEYQILLASPEVNGFFGAKGVAGAIPAAYVHIERQFFSEVCSLGQQERVQLQEYWRRGWTFHAKGLWLYLAGSSLPCLTLIGSPNFGYRSVHRDLEAQIAIVTENQALQQQLHQEQEQLYLRSGVVSSATFEQPSRQVKLWVKMVTPLIKNFF | |
RUO | |
PGS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |