Läs mer
Human Phospho-VEGFR2/KDR/Flk-1 (Y1214) Antibody, R&D Systems™
Produktkod. 18030012
Change view
Kvantitet:
50 μg
Produktkod. Kvantitet
18030012 50 μg
1 options
Produktkod. 18030012 Leverantör R&D Systems Leverantörsnummer AF1766
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Rabbit Polyclonal Antibody has been used in 5 publications
VEGFR2/KDR/Flk-1 Polyclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen VEGFR2/KDR/Flk-1 (p tyr1214)
Användningsområden Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Klassificering Polyclonal
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Gene Alias CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
Gensymboler KDR
Värd art Rabbit
Immunogen Phosphopeptide containing human VEGFR2/KDR/Flk-1 Y1214 site
Reningsmetod Affinity Purified
Kvantitet 50 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 3791
Testspecificitet Detects human VEGFR2/KDR/Flk-1 in Western blots when phosphorylated at Y1214.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Innehåll och lagring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotyp IgG