missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PIB5PA Partial ORF (NP_055237, 361 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027124-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135716

  • 6845.00 SEK / 25µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: FLLQFAFRDDMPLVRLEVADEWVRPEQAVVRYRMETVFARSSWDWIGLYRVGFRHCKDYVAYVWAKHEDVDGNTYQVTFSEESLPKGHGDFILGYYSHNHSILIGITEPF
Specifications

Specifications

NP_055237
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FLLQFAFRDDMPLVRLEVADEWVRPEQAVVRYRMETVFARSSWDWIGLYRVGFRHCKDYVAYVWAKHEDVDGNTYQVTFSEESLPKGHGDFILGYYSHNHSILIGITEPF
RUO
INPP5J
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27124
PIB5PA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
INPP5/MGC129984/PIB5PA/PIPP
INPP5J
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers