missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PIGF Partial ORF (NP_775097.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_775097.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5281 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172785
|
Abnova™
H00005281-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182785
|
Abnova™
H00005281-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor, a glycolipid containing three mannose molecules in its core backbone, is found on many blood cells where it serves to anchor proteins to the cell surface. The encoded protein and another GPI synthesis protein, PIGO, function in the transfer of ethanolaminephosphate to the third mannose in GPI. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGFVTAVNLVLYLVVKPNTSSKRSSLSHKVTGFLKCCIYFLMSCFSFHVIFVLSpecifications
NP_775097.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC32646/MGC33136 | |
PIGF | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5281 | |
PIGF (Human) Recombinant Protein (Q01) | |
MKDNDIKRLLYTHLLCIFSIILSVFIPSLFLENFSILETHLTWLCICSGFVTAVNLVLYLVVKPNTSSKRSSLSHKVTGFLKCCIYFLMSCFSFHVIFVL | |
RUO | |
PIGF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |