missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PIK4CB Partial ORF (NP_002642.1, 731 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005298-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: GGLDGDMFNYYKMLMLQGLIAARKHMDKVVQIVEIMQQGSQLPCFHGSSTIRNLKERFHMSMTEEQLQLLVEQMVDGSMRSITTKLYDGFQYLTNGIMSpecifications
NP_002642.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GGLDGDMFNYYKMLMLQGLIAARKHMDKVVQIVEIMQQGSQLPCFHGSSTIRNLKERFHMSMTEEQLQLLVEQMVDGSMRSITTKLYDGFQYLTNGIM | |
RUO | |
PI4KB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5298 | |
PIK4CB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PI4K-BETA/PI4KIIIbeta/PI4Kbeta/PIK4CB/pi4K92 | |
PI4KB | |
Recombinant | |
wheat germ expression system |