Läs mer
Human PILR-alpha Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB64842-100
This item is not returnable.
View return policy
Description
PILR-alpha Monoclonal specifically detects PILR-alpha in Human samples. It is validated for Flow Cytometry, Neutralization.
Specifications
| PILR-alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cell surface receptor FDF03, FDF03, inhibitory receptor PILRalpha, Inhibitory receptor PILR-alpha, paired immunoglobin-like receptor alpha, paired immunoglobin-like type 2 receptor alpha, paired immunoglobulin-like receptor alpha, paired immunoglobulin-like type 2 receptor alpha, PILRA, PILRalpha | |
| Mouse myeloma cell line NS0-derived recombinant human PILR-alpha with a C-terminal 6-His tag Gln20-Thr196 Accession # Q9UKJ1 | |
| 100 μg | |
| Primary | |
| Detects human PILR-alpha in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization | |
| 2175B | |
| Flow Cytometry 0.25 ug/10^6 cells, Neutralization 1-5 ug/mL | |
| Q9UKJ1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 29992 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |