missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PIP5K1B Full-length ORF (AAH30587.1, 1 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008395-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16114818

  • 6150.00 SEK / 25µg
Estimated Shipment: 12-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSSAAENGEAAPGKQNEEKTYKKTQVKNQILSRLPKIPCTFIQAWTNTEMITLLACYYFRSVSTASSAIKGAIQLGIGYTVGNLTSKPERDVLMQDFYVVESVFLPSEGSNLTPAHHYPDFRFKTYAPLAFRYFRELFGIKPDDYLYSICSEPLIELSNPGASGSLFFVTSDDEFIIKTVQHKEAEFLQKLLPGYYMNLNQNPRTLLPKFYGLYCMQSGGINIRIVVMNNVLPRSMRMHFTYDLKGSTYKRRASRKEREKSNPTFKDLDFLQDMHEGLYFDTETYNALMKTLQRDCRVLESFKIMDYSLLLGIHFLDHSLKEKEEETPQNVPDAKRTGMQKVLYSTAMESIQGPGKSGDGIITENPDTMGGIPAKSHRGEKLLLFTGIIDILQSYRLMKKLEHSWKALVYDGDTVSVHRPSFYADRFLKFMNSRVFKKIQALKASPSKKRCNSIAALKATSQEIVSSISQEWKDEKRDLLTEGQSFSSLDEEALGSRHRPDLVPSTPSLFEAASLATTISSSSLYVNEHYPHDRPTLYSNRFKMATSEH
Specifications

Specifications

AAH30587.1
Liquid
8395
PIP5K1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSAAENGEAAPGKQNEEKTYKKTQVKNQILSRLPKIPCTFIQAWTNTEMITLLACYYFRSVSTASSAIKGAIQLGIGYTVGNLTSKPERDVLMQDFYVVESVFLPSEGSNLTPAHHYPDFRFKTYAPLAFRYFRELFGIKPDDYLYSICSEPLIELSNPGASGSLFFVTSDDEFIIKTVQHKEAEFLQKLLPGYYMNLNQNPRTLLPKFYGLYCMQSGGINIRIVVMNNVLPRSMRMHFTYDLKGSTYKRRASRKEREKSNPTFKDLDFLQDMHEGLYFDTETYNALMKTLQRDCRVLESFKIMDYSLLLGIHFLDHSLKEKEEETPQNVPDAKRTGMQKVLYSTAMESIQGPGKSGDGIITENPDTMGGIPAKSHRGEKLLLFTGIIDILQSYRLMKKLEHSWKALVYDGDTVSVHRPSFYADRFLKFMNSRVFKKIQALKASPSKKRCNSIAALKATSQEIVSSISQEWKDEKRDLLTEGQSFSSLDEEALGSRHRPDLVPSTPSLFEAASLATTISSSSLYVNEHYPHDRPTLYSNRFKMATSEH
RUO
PIP5K1B
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
88.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MSS4/STM7
PIP5K1B
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers