missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PISD Partial ORF (NP_055153, 62 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_055153 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23761 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16177672
|
Abnova™
H00023761-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16167672
|
Abnova™
H00023761-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Phosphatidylserine decarboxylases (PSDs; EC 4.1.1.65) catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits (Schuiki and Daum, 2009 [PubMed 19165886]).[supplied by OMIM]
Sequence: PLSRVGWRPVSRVALYKSVPTRLLSRAWGRLNQVELPHWLRRPVYSLYIWTFGVNMKEAAVEDLHHYRNLSEFFRRKLKPQARPVCGLHSVISPSDGRILNFGQVKNCESpecifications
NP_055153 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJ858B16/DKFZp566G2246/PSDC/PSSC/dJ858B16.2 | |
PISD | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23761 | |
PISD (Human) Recombinant Protein (Q01) | |
PLSRVGWRPVSRVALYKSVPTRLLSRAWGRLNQVELPHWLRRPVYSLYIWTFGVNMKEAAVEDLHHYRNLSEFFRRKLKPQARPVCGLHSVISPSDGRILNFGQVKNCE | |
RUO | |
PISD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |