missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PITPNB Partial ORF (NP_036531, 181 a.a. - 271 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4055.00 SEK - 6150.00 SEK
Specifications
Name | PITPNB (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 35.75kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115036
|
Abnova™
H00023760-Q01.25UG |
25 ug |
6150.00 SEK
25µg |
Estimated Shipment: 18-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16105036
|
Abnova™
H00023760-Q01.10UG |
10 ug |
4055.00 SEK
10µg |
Estimated Shipment: 18-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
The protein encoded by this gene is found in the cytoplasm, where it catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes. [provided by RefSeq]
Sequence: LANSPDCPQMCAYKLVTIKFKWWGLQSKVENFIQKQEKRIFTNFHRQLFCWIDKWIDLTMEDIRRMEDETQKELETMRKRGSVRGTSAADVSpecifications
PITPNB (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
35.75kDa |