missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PKD2L1 Full-length ORF (NP_057196.2, 1 a.a. - 805 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_057196.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9033 |
Molecular Weight (g/mol) | 118.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173202
|
Abnova™
H00009033-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16163202
|
Abnova™
H00009033-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. This protein functions as a calcium-regulated nonselective cation channel. Alternative splice variants have been described but their full length sequences have not been determined. [provided by RefSeq]
Sequence: MNAVGSPEGQELQKLGSGAWDNPAYSGPPSPHGTLRVCTISSTGPLQPQPKKPEDEPQETAYRTQVSSCCLHICQGIRGLWGTTLTENTAENRELYIKTTLRELLVYIVFLVDICLLTYGMTSSSAYYYTKVMSELFLHTPSDTGVSFQAISSMADFWDFAQGPLLDSLYWTKWYNNQSLGHGSHSFIYYENMLLGVPRLRQLKVRNDSCVVHEDFREDILSCYDVYSPDKEEQLPFGPFNGTAWTYHSQDELGGFSHWGRLTSYSGGGYYLDLPGSRQGSAEALRALQEGLWLDRGTRVVFIDFSVYNANINLFCVLRLVVEFPATGGAIPSWQIRTVKLIRYVSNWDFFIVGCEVIFCVFIFYYVVEEILELHIHRLRYLSSIWNILDLVVILLSIVAVGFHIFRTLEVNRLMGKLLQQPNTYADFEFLAFWQTQYNNMNAVNLFFAWIKIFKYISFNKTMTQLSSTLARCAKDILGFAVMFFIVFFAYAQLGYLLFGTQVENFSTFIKCIFTQFRIILGDFDYNAIDNANRILGPAYFVTYVFFVFFVLLNMFLAIINDTYSEVKEELAGQKDELQLSDLLKQGYNKTLLRLRLRKERVSDVQKVLQGGEQEIQFEDFTNTLRELGHAEHEITELTATFTKFDRDGNRILDEKEQEKMRQDLEEERVALNTEIEKLGRSIVSSPQGKSGPEAARAGGWVSGEEFYMLTRRVLQLETVLEGVVSQIDAVGSKLKMLERKGWLAPSPGVKEQAIWKHPQPAPAVTPDPWGVQGGQESEVPYKREEEALEERRLSRGEIPTLQRSSpecifications
NP_057196.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNAVGSPEGQELQKLGSGAWDNPAYSGPPSPHGTLRVCTISSTGPLQPQPKKPEDEPQETAYRTQVSSCCLHICQGIRGLWGTTLTENTAENRELYIKTTLRELLVYIVFLVDICLLTYGMTSSSAYYYTKVMSELFLHTPSDTGVSFQAISSMADFWDFAQGPLLDSLYWTKWYNNQSLGHGSHSFIYYENMLLGVPRLRQLKVRNDSCVVHEDFREDILSCYDVYSPDKEEQLPFGPFNGTAWTYHSQDELGGFSHWGRLTSYSGGGYYLDLPGSRQGSAEALRALQEGLWLDRGTRVVFIDFSVYNANINLFCVLRLVVEFPATGGAIPSWQIRTVKLIRYVSNWDFFIVGCEVIFCVFIFYYVVEEILELHIHRLRYLSSIWNILDLVVILLSIVAVGFHIFRTLEVNRLMGKLLQQPNTYADFEFLAFWQTQYNNMNAVNLFFAWIKIFKYISFNKTMTQLSSTLARCAKDILGFAVMFFIVFFAYAQLGYLLFGTQVENFSTFIKCIFTQFRIILGDFDYNAIDNANRILGPAYFVTYVFFVFFVLLNMFLAIINDTYSEVKEELAGQKDELQLSDLLKQGYNKTLLRLRLRKERVSDVQKVLQGGEQEIQFEDFTNTLRELGHAEHEITELTATFTKFDRDGNRILDEKEQEKMRQDLEEERVALNTEIEKLGRSIVSSPQGKSGPEAARAGGWVSGEEFYMLTRRVLQLETVLEGVVSQIDAVGSKLKMLERKGWLAPSPGVKEQAIWKHPQPAPAVTPDPWGVQGGQESEVPYKREEEALEERRLSRGEIPTLQRS | |
RUO | |
PKD2L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9033 | |
PKD2L1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCL/PKD2L/PKDL | |
PKD2L1 | |
Recombinant | |
wheat germ expression system |