missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PLA2G12B Partial ORF (NP_115951.2, 78 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
POA
Specifications
Name | PLA2G12B (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
Molecular Weight (g/mol) | 37.84kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
Phospholipase A2 (PLA2) enzymes catalyze hydrolysis of glycolipids to release free fatty acids and lysophospholipids. PLA2G12B belongs to the PLA2 family, but it is catalytically inactive due to an amino acid change in its active site and has altered phospholipid-binding properties (Rouault et al., 2003 [PubMed 14516201]).[supplied by OMIM]
Sequence: PGYKPQEPNGCGSYFLGLKVPESMDLGIPAMTKCCNQLDVCYDTCGANKYRCDAKFRWCLHSICSDLKRSLGFVSKVEAACDSLVDTVFNTVWTLGCRPFMNSQRAACICSpecifications
PLA2G12B (Human) Recombinant Protein (Q01) | |
Yes | |
37.84kDa | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |