missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLEKHA1 Partial ORF (NP_001001974.1, 118 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00059338-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179366

  • 5030.41 SEK / 25µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: QSDSQPNSDNLSRHGECGKKQVSYRTDIVGGVPIITPTQKEEVNECGESIDRNNLKRSQSHLPYFTPKPPQDSAVIKAGYCVKQGAVMKNWKRRYFQLD
Specifications

Specifications

NP_001001974.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QSDSQPNSDNLSRHGECGKKQVSYRTDIVGGVPIITPTQKEEVNECGESIDRNNLKRSQSHLPYFTPKPPQDSAVIKAGYCVKQGAVMKNWKRRYFQLD
RUO
PLEKHA1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
59338
PLEKHA1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TAPP1
PLEKHA1
Yes
Videos
SDS
Documents
Certificates