missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLEKHM1 Partial ORF (AAH64361, 957 a.a. - 1056 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009842-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16171356

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is essential for bone resorption, and may play a critical role in vesicular transport in the osteoclast. Mutations in this gene are associated with autosomal recessive osteopetrosis type 6 (OPTB6). Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Sequence: PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFA
Specifications

Specifications

AAH64361
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFA
RUO
PLEKHM1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9842
PLEKHM1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AP162/B2/KIAA0356/OPTB6
PLEKHM1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers