Learn More
Abnova™ Human PLEKHM1 Partial ORF (AAH64361, 957 a.a. - 1056 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009842-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is essential for bone resorption, and may play a critical role in vesicular transport in the osteoclast. Mutations in this gene are associated with autosomal recessive osteopetrosis type 6 (OPTB6). Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Sequence: PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFASpecifications
AAH64361 | |
Liquid | |
9842 | |
PLEKHM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP162/B2/KIAA0356/OPTB6 | |
PLEKHM1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PHRFSVADLQQIADGVYEGFLKALIEFASQHVYHCDLCTQRGFICQICQHHDIIFPFEFDTTVRCAECKTVFHQSCQAVVKKGCPRCARRRKYQEQNIFA | |
RUO | |
PLEKHM1 | |
Wheat Germ (in vitro) | |
GST |