Learn More
Abnova™ Human PMP22 Full-length ORF (NP_000295.1, 1 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005376-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an integral membrane protein that is a major component of myelin in the peripheral nervous system. Various mutations of this gene are causes of Charcot-Marie-Tooth disease Type IA, Dejerine-Sottas syndrome, and hereditary neuropathy with liability to pressure palsies. Alternative splicing of this gene results in three transcript variants that encode the same protein. [provided by RefSeq]
Sequence: MLLLLLSIIVLHVAVLVLLFVSTIVSQWIVGNGHATDLWQNCSTSSSGNVHHCFSSSPNEWLQSVQATMILSIIFSILSLFLFFCQLFTLTKGGRFYITGIFQILAGLCVMSAAAIYTVRHPEWHLNSDYSYGFAYILAWVAFPLALLSGVIYVILRKRESpecifications
NP_000295.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMT1A/CMT1E/DSS/GAS-3/HMSNIA/HNPP/MGC20769/Sp110 | |
PMP22 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5376 | |
PMP22 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLLLLLSIIVLHVAVLVLLFVSTIVSQWIVGNGHATDLWQNCSTSSSGNVHHCFSSSPNEWLQSVQATMILSIIFSILSLFLFFCQLFTLTKGGRFYITGIFQILAGLCVMSAAAIYTVRHPEWHLNSDYSYGFAYILAWVAFPLALLSGVIYVILRKRE | |
RUO | |
PMP22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |