Learn More
Abnova™ Human POLD2 Full-length ORF (NP_006221.1, 1 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005425-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The DNA polymerase delta complex is involved in DNA replication and repair, and it consists of the proliferating cell nuclear antigen (PCNA; MIM 176740), the multisubunit replication factor C (see MIM 102579), and the 4 subunit polymerase complex: POLD1 (MIM 174761), POLD2, POLD3 (MIM 611415), and POLD4 (MIM 611525) (Liu and Warbrick, 2006 [PubMed 16934752]).[supplied by OMIM]
Sequence: MFSEQAAQRAHTLLSPPSANNATFARVPVATYTNSSQPFRLGERSFSRQYAHIYATRLIQMRPFLENRAQQHWGSGVGVKKLCELQPEEKCCVVGTLFKAMPLQPSILREVSEEHNLLPQPPRSKYIHPDDELVLEDELQRIKLKGTIDVSKLVTGTVLAVFGSVRDDGKFLVEDYCFADLAPQKPAPPLDTDRFVLLVSGLGLGGGGGESLLGTQLLVDVVTGQLGDEGEQCSAAHVSRVILAGNLLSHSTQSRDSINKAKYLTKKTQAASVEAVKMLDEILLQLSASVPVDVMPGEFDPTNYTLPQQPLHPCMFPLATAYSTLQLVTNPYQATIDGVRFLGTSGQNVSDIFRYSSMEDHLEILEWTLRVRHISPTAPDTLGCYPFYKTDPFIFPECPHVYFCGNTPSFGSKIIRGPEDQTVLLVTVPDFSATQTACLVNLRSLACQPISFSGFGAEDDDLGGLGLGPSpecifications
NP_006221.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POLD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5425 | |
POLD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFSEQAAQRAHTLLSPPSANNATFARVPVATYTNSSQPFRLGERSFSRQYAHIYATRLIQMRPFLENRAQQHWGSGVGVKKLCELQPEEKCCVVGTLFKAMPLQPSILREVSEEHNLLPQPPRSKYIHPDDELVLEDELQRIKLKGTIDVSKLVTGTVLAVFGSVRDDGKFLVEDYCFADLAPQKPAPPLDTDRFVLLVSGLGLGGGGGESLLGTQLLVDVVTGQLGDEGEQCSAAHVSRVILAGNLLSHSTQSRDSINKAKYLTKKTQAASVEAVKMLDEILLQLSASVPVDVMPGEFDPTNYTLPQQPLHPCMFPLATAYSTLQLVTNPYQATIDGVRFLGTSGQNVSDIFRYSSMEDHLEILEWTLRVRHISPTAPDTLGCYPFYKTDPFIFPECPHVYFCGNTPSFGSKIIRGPEDQTVLLVTVPDFSATQTACLVNLRSLACQPISFSGFGAEDDDLGGLGLGP | |
RUO | |
POLD2 | |
Recombinant | |
wheat germ expression system |