missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human POLE4 Partial ORF (NP_063949, 31 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056655-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16182233

  • 5030.41 SEK / 25µg

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

POLE4 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.[supplied by OMIM]

Sequence: TSVPGARLSRLPLARVKALVKADPDVTLAGQEAIFILARAAELFVETIAKDAYCCAQQGKRKTLQRRDLDNAIEAVDEFAFLEGTLD
Specifications

Specifications

NP_063949
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.31kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TSVPGARLSRLPLARVKALVKADPDVTLAGQEAIFILARAAELFVETIAKDAYCCAQQGKRKTLQRRDLDNAIEAVDEFAFLEGTLD
RUO
POLE4
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
56655
POLE4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
p12
POLE4
Yes
Videos
SDS
Documents
Certificates