missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human PP14/Glycodelin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF842
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PP14/Glycodelin Polyclonal specifically detects PP14/Glycodelin in Human samples. It is validated for Western Blot.
Specifikationer
| PP14/Glycodelin | |
| Polyclonal | |
| Biotin | |
| P09466 | |
| PAEP | |
| Mouse myeloma cell line NS0-derived recombinant human PP14/Glycodelin Met19-Phe180 (Glu95Gly) Accession # P09466 | |
| 50 μg | |
| Primary | |
| Detects human PP14/Glycodelin in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PAEP, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5047 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering