missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PPFIA3 Partial ORF (NP_003651, 1043 a.a. - 1139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_003651 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8541 |
Molecular Weight (g/mol) | 36.41kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138775
|
Abnova™
H00008541-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128775
|
Abnova™
H00008541-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family. Liprins interact with members of LAR family of transmembrane protein tyrosine phosphatases, which are known to be important for axon guidance and mammary gland development. Liprin family protein has been shown to localize phosphatase LAR to cell focal adhesions and may be involved in the molecular organization of presynaptic active zones. [provided by RefSeq]
Sequence: NERVMGWVSGLGLKEFATNLTESGVHGALLALDETFDYSDLALLLQIPTQNAQARQLLEKEFSNLISLGTDRRLDEDSAKSFSRSPSWRKMFREKDLSpecifications
NP_003651 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0654/LPNA3/MGC126567/MGC126569 | |
PPFIA3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8541 | |
PPFIA3 (Human) Recombinant Protein (Q01) | |
NERVMGWVSGLGLKEFATNLTESGVHGALLALDETFDYSDLALLLQIPTQNAQARQLLEKEFSNLISLGTDRRLDEDSAKSFSRSPSWRKMFREKDL | |
RUO | |
PPFIA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |