missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPHLN1 Full-length ORF (NP_958923.1, 1 a.a. - 374 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051535-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180113

  • 5030.41 SEK / 25µg

Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is one of the several proteins that become sequentially incorporated into the cornified cell envelope during the terminal differentiation of keratinocyte at the outer layers of epidermis. This protein interacts with periplakin, which is known as a precursor of the cornified cell envelope. The cellular localization pattern and insolubility of this protein suggest that it may play a role in epithelial differentiation and contribute to epidermal integrity and barrier formation. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MAYRRDEMWSEGRYEYERIPRERAPPRSHPSDGYNRLVNIVPKKPPLLDRPGEGSYNRYYSHVDYRDYDEGRSFSHDRRSGPPHRGDESGYRWTRDDHSASRQPEYRDMRDGFRRKSFYSSHYARERSPYKRDNTFFRESPVGRKDSPHSRSGSSVSSRSYSPERSKSYSFHQSQHRKSVRPGASYKRQNEGNPERDKERPVQSLKTSRDTSPSSGSAVSSSKVLDKPSRLTEKELAEAASKWAAEKLEKSDESNLPEISEYEAGSTAPLFTDQPEEPESNTTHGIELFEDSQLTTRSKAIASKTKEIEQVYRQDCETFGMVVKMLIEKDPSLEKSIQFALRQNLHEIGERCVEELKHFIAEYDTSTQDFGEPF
Specifications

Specifications

NP_958923.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSPC206/HSPC232/MGC48786
PPHLN1
Yes
wheat germ expression system
Liquid
51535
PPHLN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAYRRDEMWSEGRYEYERIPRERAPPRSHPSDGYNRLVNIVPKKPPLLDRPGEGSYNRYYSHVDYRDYDEGRSFSHDRRSGPPHRGDESGYRWTRDDHSASRQPEYRDMRDGFRRKSFYSSHYARERSPYKRDNTFFRESPVGRKDSPHSRSGSSVSSRSYSPERSKSYSFHQSQHRKSVRPGASYKRQNEGNPERDKERPVQSLKTSRDTSPSSGSAVSSSKVLDKPSRLTEKELAEAASKWAAEKLEKSDESNLPEISEYEAGSTAPLFTDQPEEPESNTTHGIELFEDSQLTTRSKAIASKTKEIEQVYRQDCETFGMVVKMLIEKDPSLEKSIQFALRQNLHEIGERCVEELKHFIAEYDTSTQDFGEPF
RUO
PPHLN1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates