missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPIL5 Partial ORF (NP_982292, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00122769-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16161647

  • 6150.00 SEK / 25µg
Estimated Shipment: 14-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene contains a leucine-rich repeat (LRR). It specifically interacts with TNFRSF9/4-1BB, a member of the tumor necrosis factor receptor (TNFR) superfamily. Overexpression of this gene suppresses the activation of NF-kappa B induced by TNFRSF9 or TNF receptor-associated factor 2 (TRAF2), which suggests that this protein is a negative regulator of TNFRSF9-mediated signaling cascades. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]

Sequence: MKLHCEVEVISRHLPALGLRNRGKGVRAVLSLCQQTSRSQPPVRAFLLISTLKDKRGTRYELRENIEQFFTKFVDEGKATVRLKEPPVDICLSKDSIWLS
Specifications

Specifications

NP_982292
Liquid
122769
PPIL5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4-1BBLRR/LRR-1/MGC20689
PPIL5
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKLHCEVEVISRHLPALGLRNRGKGVRAVLSLCQQTSRSQPPVRAFLLISTLKDKRGTRYELRENIEQFFTKFVDEGKATVRLKEPPVDICLSKDSIWLS
RUO
PPIL5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers