missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP2R2D Full-length ORF (AAH72402.1, 1 a.a. - 288 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055844-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16161873

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MDLMVEASPRRIFANAHTYHINSISVNSDHETYLSADDLRINLWHLEITDRSFNIVDIKPANMEELTEVITAAEFHPHQCNVFVYSSSKGTIRLCDMRSSALCDRHSKFFEEPEDPSSRSFFSEIISSISDVKFSHSGRYMMTRDYLSVKVWDLNMESRPVETHQVHEYLRSKLCSLYENDCIFDKFECCWNGSDSAIMTGSYNNFFRMFDRDTRRDVTLEASRESSKPRASLKPRKVCTGGKRRKDEISVDSLDFNKKILHTAWHPVDNVIAVAATNNLYIFQDKIN
Specifications

Specifications

AAH72402.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MDS026
PPP2R2D
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55844
PPP2R2D (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDLMVEASPRRIFANAHTYHINSISVNSDHETYLSADDLRINLWHLEITDRSFNIVDIKPANMEELTEVITAAEFHPHQCNVFVYSSSKGTIRLCDMRSSALCDRHSKFFEEPEDPSSRSFFSEIISSISDVKFSHSGRYMMTRDYLSVKVWDLNMESRPVETHQVHEYLRSKLCSLYENDCIFDKFECCWNGSDSAIMTGSYNNFFRMFDRDTRRDVTLEASRESSKPRASLKPRKVCTGGKRRKDEISVDSLDFNKKILHTAWHPVDNVIAVAATNNLYIFQDKIN
RUO
PPP2R2D
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers