Learn More
Abnova™ Human PRICKLE1 Partial ORF (NP_694571, 684 a.a. - 756 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00144165-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear receptor that may be a negative regulator of the Wnt/beta-catenin signaling pathway. The encoded protein localizes to the nuclear membrane and has been implicated in the nuclear trafficking of the transcription repressors REST/NRSF and REST4. Mutations in this gene have been linked to progressive myoclonus epilepsy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 3. [provided by RefSeq]
Sequence: DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYGSpecifications
NP_694571 | |
Liquid | |
144165 | |
PRICKLE1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EPM1B/FLJ31627/FLJ31937/MGC138902/MGC138903/RILP | |
PRICKLE1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DNALNLVTERKYSPKDRLRLYTPDNYEKFIQNKSAREIQAYIQNADLYGQYAHATPDYGLQNPGMNRFLGLYG | |
RUO | |
PRICKLE1 | |
Wheat Germ (in vitro) | |
GST |