Learn More
Abnova™ Human PRIM1 Partial ORF (AAH05266, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005557-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The replication of DNA in eukaryotic cells is carried out by a complex chromosomal replication apparatus, in which DNA polymerase alpha and primase are two key enzymatic components. Primase, which is a heterodimer of a small subunit and a large subunit, synthesizes small RNA primers for the Okazaki fragments made during discontinuous DNA replication. The protein encoded by this gene is the small, 49 kDa primase subunit. [provided by RefSeq]
Sequence: METFDPTELPELLKLYYRRLFPYSQYYRWLNYGGVIKNYFQHREFSFTLKDDIYIRYQSFNNQSDLEKEMQKMNPYKIDIGAVYSHRPNQHNTVKLGAFQSpecifications
AAH05266 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12308/p49 | |
PRIM1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5557 | |
PRIM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
RUO | |
PRIM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |