Learn More
Abnova™ Human PRKCDBP Full-length ORF (AAH11585.1, 1 a.a. - 261 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00112464-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene was identified as a binding protein of the protein kinase C, delta (PRKCD). The expression of this gene in cultured cell lines is strongly induced by serum starvation. The expression of this protein was found to be down-regulated in various cancer cell lines, suggesting the possible tumor suppressor function of this protein. [provided by RefSeq]
Sequence: MRESALEPGPVPEAPAGGPVHAVTVVTLLEKLASMLETLRERQGGLARRQGGLAGSVRRIQSGLGALSRSHDTTSNTLAQLLAKAERVSSHANAAQERAVRRAAQVQRLEANHGLLVARGKLHVLLFKEEGEVPASAFQKAPEPLGPADQSELGPEQLEAEVGESSDEEPVESRAQRLRRTGLQKVQSLRRALSGRKGPAAPPPTPVKPPRLGPGRSAEAQPEAQPALEPTLEPEPPQDTEEDPGRPGAAEEALLQMESVASpecifications
AAH11585.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSRBC/MGC20400/SRBC | |
PRKCDBP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
112464 | |
PRKCDBP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRESALEPGPVPEAPAGGPVHAVTVVTLLEKLASMLETLRERQGGLARRQGGLAGSVRRIQSGLGALSRSHDTTSNTLAQLLAKAERVSSHANAAQERAVRRAAQVQRLEANHGLLVARGKLHVLLFKEEGEVPASAFQKAPEPLGPADQSELGPEQLEAEVGESSDEEPVESRAQRLRRTGLQKVQSLRRALSGRKGPAAPPPTPVKPPRLGPGRSAEAQPEAQPALEPTLEPEPPQDTEEDPGRPGAAEEALLQMESVA | |
RUO | |
PRKCDBP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |