missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRKY Full-length ORF (NP_002751.1, 1 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005616-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119671

  • 6150.00 SEK / 25µg
Estimated Shipment: 12-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is similar to the protein kinase, X-linked gene in the pseudoautosomal region of the X chromsoome. The gene is classified as a transcribed pseudogene because it has lost a coding exon that results in all transcripts being candidates for nonsense-mediated decay (NMD) and unlikely to express a protein. Abnormal recombination between this gene and a related gene on chromosome X is a frequent cause of XX males and XY females. [provided by RefSeq]

Sequence: MEAPGPAQAAAAESNSREVTEDAADWAPALCPSPEARSPEAPAYRLQDCDALVTMGTGTFGRVHLVKEKTAKHFFALKVMSIPDVIRRKQEQHVHNEKSVLKEVSHPFLIRLFWTWHEERFLYMLMEYVPGGELFSYLRNRGHFSSTTGLFYSAEIICAIEYLHSKEIVYRDLKPENILLDRDGHIKLTDFGFAKKLVDRTWTLCGTPEYLAPEVIQSKGHGRAVDWWALGILIFEMLSGFPPFFDDNPFGIYQKILAGKLYFPRHLDFHVKTGRMM
Specifications

Specifications

NP_002751.1
Liquid
5616
PRKY (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEAPGPAQAAAAESNSREVTEDAADWAPALCPSPEARSPEAPAYRLQDCDALVTMGTGTFGRVHLVKEKTAKHFFALKVMSIPDVIRRKQEQHVHNEKSVLKEVSHPFLIRLFWTWHEERFLYMLMEYVPGGELFSYLRNRGHFSSTTGLFYSAEIICAIEYLHSKEIVYRDLKPENILLDRDGHIKLTDFGFAKKLVDRTWTLCGTPEYLAPEVIQSKGHGRAVDWWALGILIFEMLSGFPPFFDDNPFGIYQKILAGKLYFPRHLDFHVKTGRMM
RUO
PRKY
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRKY
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers