missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRRX1 Full-length ORF (NP_008833.1, 1 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005396-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139261

  • 6150.00 SEK / 25µg
Estimated Shipment: 05-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription co-activator, enhancing the DNA-binding activity of serum response factor, a protein required for the induction of genes by growth and differentiation factors. The protein regulates muscle creatine kinase, indicating a role in the establishment of diverse mesodermal muscle types. Alternative splicing yields two isoforms that differ in abundance and expression patterns. [provided by RefSeq]

Sequence: MTSSYGHVLERQPALGGRLDSPGNLDTLQAKKNFSVSHLLDLEEAGDMVAAQADENVGEAGRSLLESPGLTSGSDTPQQDNDQLNSEEKKKRKQRRNRTTFNSSQLQALERVFERTHYPDAFVREDLARRVNLTEARVQVWFQNRRAKFRRNERAMLANKNASLLKSYSGDVTAVEQPIVPRPAPRPTDYLSWGTASPYRSSSLPRCCLHEGLHNGF
Specifications

Specifications

NP_008833.1
Liquid
5396
PRRX1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSSYGHVLERQPALGGRLDSPGNLDTLQAKKNFSVSHLLDLEEAGDMVAAQADENVGEAGRSLLESPGLTSGSDTPQQDNDQLNSEEKKKRKQRRNRTTFNSSQLQALERVFERTHYPDAFVREDLARRVNLTEARVQVWFQNRRAKFRRNERAMLANKNASLLKSYSGDVTAVEQPIVPRPAPRPTDYLSWGTASPYRSSSLPRCCLHEGLHNGF
RUO
PRRX1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PHOX1/PMX1/PRX1
PRRX1
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers