Learn More
Abnova™ Human PRSS12 Partial ORF (NP_003610.1, 541 a.a. - 640 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008492-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the trypsin family of serine proteases. Studies in mouse suggest that the encoded enzyme may be involved in structural reorganizations associated with learning and memory. The enzyme is also expressed in Leydig cells in the testis, but its function in this tissue is unknown. [provided by RefSeq]
Sequence: LGYKGPARARTMAYFGEGKGPIHVDNVKCTGNERSLADCIKQDIGRHNCRHSEDAGVICDYFGKKASGNSNKESLSSVCGLRLLHRRQKRIIGGKNSLRGSpecifications
NP_003610.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGYKGPARARTMAYFGEGKGPIHVDNVKCTGNERSLADCIKQDIGRHNCRHSEDAGVICDYFGKKASGNSNKESLSSVCGLRLLHRRQKRIIGGKNSLRG | |
RUO | |
PRSS12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8492 | |
PRSS12 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BSSP-3/BSSP3/MGC12722/MRT1 | |
PRSS12 | |
Recombinant | |
wheat germ expression system |