Learn More
Abnova™ Human PSMD5 Full-length ORF (NP_005038.1, 1 a.a. - 504 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005038.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5711 |
Molecular Weight (g/mol) | 82.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141918
|
Abnova™
H00005711-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131918
|
Abnova™
H00005711-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator base. [provided by RefSeq]
Sequence: MAAQALALLREVARLEAPLEELRALHSVLQAVPLNELRQQAAELRLGPLFSLLNENHREKTTLCVSILERLLQAMEPVHVARNLRVDLQRGLIHPDDSVKILTLSQIGRIVENSDAVTEILNNAELLKQIVYCIGGENLSVAKAAIKSLSRISLTQAGLEALFESNLLDDLKSVMKTNDIVRYRVYELIIEISSVSPESLNYCTTSGLVTQLLRELTGEDVLVRATCIEMVTSLAYTHHGRQYLAQEGVIDQISNIIVGADSDPFSSFYLPGFVKFFGNLAVMDSPQQICERYPIFVEKVFEMIESQDPTMIGVAVDTVGILGSNVEGKQVLQKTGTRFERLLMRIGHQSKNAPVELKIRCLDAISSLLYLPPEQQTDDLLRMTESWFSSLSRDPLELFRGISSQPFPELHCAALKVFTAIANQPWAQKLMFNSPGFVEYVVDRSVEHDKASKDAKYELVKALANSKTIAEIFGNPNYLRLRTYLSEGPYYVKPVSTTAVEGAESpecifications
NP_005038.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAQALALLREVARLEAPLEELRALHSVLQAVPLNELRQQAAELRLGPLFSLLNENHREKTTLCVSILERLLQAMEPVHVARNLRVDLQRGLIHPDDSVKILTLSQIGRIVENSDAVTEILNNAELLKQIVYCIGGENLSVAKAAIKSLSRISLTQAGLEALFESNLLDDLKSVMKTNDIVRYRVYELIIEISSVSPESLNYCTTSGLVTQLLRELTGEDVLVRATCIEMVTSLAYTHHGRQYLAQEGVIDQISNIIVGADSDPFSSFYLPGFVKFFGNLAVMDSPQQICERYPIFVEKVFEMIESQDPTMIGVAVDTVGILGSNVEGKQVLQKTGTRFERLLMRIGHQSKNAPVELKIRCLDAISSLLYLPPEQQTDDLLRMTESWFSSLSRDPLELFRGISSQPFPELHCAALKVFTAIANQPWAQKLMFNSPGFVEYVVDRSVEHDKASKDAKYELVKALANSKTIAEIFGNPNYLRLRTYLSEGPYYVKPVSTTAVEGAE | |
RUO | |
PSMD5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5711 | |
PSMD5 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0072/MGC23145/S5B | |
PSMD5 | |
Recombinant | |
wheat germ expression system |