missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTPN12 Full-length ORF (NP_002826.2, 1 a.a. - 780 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005782-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179901

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains a C-terminal PEST motif, which serves as a protein-protein interaction domain, and may regulate protein intracellular half-life. This PTP was found to bind and dephosphorylate the product of the oncogene c-ABL and thus may play a role in oncogenesis. This PTP was also shown to interact with, and dephosphorylate, various products related to cytoskeletal structure and cell adhesion, such as p130 (Cas), CAKbeta/PTK2B, PSTPIP1, and paxillin. This suggests it has a regulatory role in controlling cell shape and mobility. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MEQVEILRKFIQRVQAMKSPDHNGEDNFARDFMRLRRLSTKYRTEKIYPTATGEKEENVKKNRYKDILPFDHSRVKLTLKTPSQDSDYINANFIKGVYGPKAYVATQGPLANTVIDFWRMIWEYNVVIIVMACREFEMGRKKCERYWPLYGEDPITFAPFKISCEDEQARTDYFIRTLLLEFQNESRRLYQFHYVNWPDHDVPSSFDSILDMISLMRKYQEHEDVPICIHCSAGCGRTGAICAIDYTWNLLKAGKIPEEFNVFNLIQEMRTQRHSAVQTKEQYELVHRAIAQLFEKQLQLYEIHGAQKIADGVNEINTENMISSIEPEKQDSPPPKPPRTRSCLVEGDAKEEILQPPEPHPVPPILTPSPPSAFPTVTTVWQDNDRYHPKPVLHMVSSEQHSADLNRNYSKSTELPGKNESTIEQIDKKLERNLSFEIKKVPLQEGPKSFDGNTLLNRGHAIKIKSASPCIADKISKPQELSSDLNVGDTSQNSCVDCSVTQSNKVSVTPPEESQNSDTPPRPDRLPLDEKGHVTWSFHGPENAIPIPDLSEGNSSDINYQTRKTVSLTPSPTTQVETPDLVDHDNTSPLFRTPLSFTNPLHSDDSDSDERNSDGAVTQNKTNISTASATVSAATSTESISTRKVLPMSIARHNIAGTTHSGAEKDVDVSEDSPPPLPERTPESFVLASEHNTPVRSEWSELQSQERSEQKKSEGLITSENEKCDHPAGGIHYEMCIECPPTFSDKREQISENPTEATDIGFGNRCGKPKGPRDPPSEWT
Specifications

Specifications

NP_002826.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PTP-PEST/PTPG1
PTPN12
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5782
PTPN12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEQVEILRKFIQRVQAMKSPDHNGEDNFARDFMRLRRLSTKYRTEKIYPTATGEKEENVKKNRYKDILPFDHSRVKLTLKTPSQDSDYINANFIKGVYGPKAYVATQGPLANTVIDFWRMIWEYNVVIIVMACREFEMGRKKCERYWPLYGEDPITFAPFKISCEDEQARTDYFIRTLLLEFQNESRRLYQFHYVNWPDHDVPSSFDSILDMISLMRKYQEHEDVPICIHCSAGCGRTGAICAIDYTWNLLKAGKIPEEFNVFNLIQEMRTQRHSAVQTKEQYELVHRAIAQLFEKQLQLYEIHGAQKIADGVNEINTENMISSIEPEKQDSPPPKPPRTRSCLVEGDAKEEILQPPEPHPVPPILTPSPPSAFPTVTTVWQDNDRYHPKPVLHMVSSEQHSADLNRNYSKSTELPGKNESTIEQIDKKLERNLSFEIKKVPLQEGPKSFDGNTLLNRGHAIKIKSASPCIADKISKPQELSSDLNVGDTSQNSCVDCSVTQSNKVSVTPPEESQNSDTPPRPDRLPLDEKGHVTWSFHGPENAIPIPDLSEGNSSDINYQTRKTVSLTPSPTTQVETPDLVDHDNTSPLFRTPLSFTNPLHSDDSDSDERNSDGAVTQNKTNISTASATVSAATSTESISTRKVLPMSIARHNIAGTTHSGAEKDVDVSEDSPPPLPERTPESFVLASEHNTPVRSEWSELQSQERSEQKKSEGLITSENEKCDHPAGGIHYEMCIECPPTFSDKREQISENPTEATDIGFGNRCGKPKGPRDPPSEWT
RUO
PTPN12
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers