Learn More
Abnova™ Human PTPRG Full-length ORF (AAH47734.1, 1 a.a. - 102 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005793-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem intracytoplasmic catalytic domains, and thus represents a receptor-type PTP. The extracellular region of this PTP contains a carbonic anhydrase-like (CAH) domain, which is also found in the extracellular region of PTPRBETA/ZETA. This gene is located in a chromosomal region that is frequently deleted in renal cell carcinoma and lung carcinoma, thus is thought to be a candidate tumor suppressor gene. [provided by RefSeq]
Sequence: MRRLLEPCWWILFLKITSSVLHYVVCFPALTEGYVGALHENRHGSAVQIRRRKASGDPYWAYSGKSSCSNGDHTGQLTSHSTHLKAHFESLLCLSSVCSNLSSpecifications
AAH47734.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HPTPG/PTPG/R-PTP-GAMMA/RPTPG | |
PTPRG | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5793 | |
PTPRG (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRLLEPCWWILFLKITSSVLHYVVCFPALTEGYVGALHENRHGSAVQIRRRKASGDPYWAYSGKSSCSNGDHTGQLTSHSTHLKAHFESLLCLSSVCSNLS | |
RUO | |
PTPRG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |